ENA311759356 Enamine

2-(4-bromophenyl)-2-chloroacetamide

  • MDL number MFCD02077565
  • Cas Number : 315232-09-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164...166
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70259

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70259
Availability details for
Quantity
  • Availability
  • Add to Cart