ENA311759360 Enamine

N1-[(3S)-2-oxooxolan-3-yl]benzene-1,4-disulfonamide

  • MDL number MFCD17977036
  • Cas Number : 1258701-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-70263

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70263
Availability details for
Quantity
  • Availability
  • Add to Cart