ENA311759362 Enamine

4H,6H,7H-thieno[3,2-c]pyran-4-ylmethanamine

  • MDL number MFCD08690319
  • Cas Number : 63932-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55804

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55804
Availability details for
Quantity
  • Availability
  • Add to Cart