ENA311759370 Enamine

(1H-1,2,3-benzotriazol-5-yl)methanol

  • MDL number MFCD00963121
  • Cas Number : 106429-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-150
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70284

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70284
Availability details for
Quantity
  • Availability
  • Add to Cart