ENA311759371 Enamine

4-(2,5-difluorobenzoyl)pyridine

  • MDL number MFCD11210309
  • Cas Number : 150020-06-7
  • Molecular Weight : 219.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70285
Availability details for
Quantity
  • Availability
  • Add to Cart