ENA311759372 Enamine

2-[cyclopentyl(methyl)amino]acetamide

  • MDL number MFCD11775756
  • Cas Number : 1090870-04-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70286

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70286
Availability details for
Quantity
  • Availability
  • Add to Cart