ENA311759373 Enamine

N-(1-cyanocyclopentyl)-2-methylpropanamide

  • MDL number MFCD12783693
  • Cas Number : 1250062-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70287

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70287
Availability details for
Quantity
  • Availability
  • Add to Cart