ENA311759379 Enamine

3-(cyclopropylsulfamoyl)-2,6-difluorobenzoic acid

  • MDL number MFCD11522359
  • Cas Number : 1039836-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70321

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70321
Availability details for
Quantity
  • Availability
  • Add to Cart