ENA311759383 Enamine

4-(difluoromethoxy)-1,3-benzothiazol-2-amine

  • MDL number MFCD11189737
  • Cas Number : 1038981-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70325

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70325
Availability details for
Quantity
  • Availability
  • Add to Cart