ENA311759388 Enamine

N-(2-aminoethyl)benzenesulfonamide

  • MDL number MFCD01310608
  • Cas Number : 42988-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74...76
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57582

Safety Information

msds https://www.enaminestore.com/catalog/EN300-57582
Availability details for
Quantity
  • Availability
  • Add to Cart