ENA311759390 Enamine

4-methyl-5-(piperidin-4-yl)-4H-1,2,4-triazole-3-thiol hydrochloride

  • MDL number MFCD15200707
  • Cas Number : 1158193-23-7
  • Molecular Weight : 234.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70332

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70332
Availability details for
Quantity
  • Availability
  • Add to Cart