ENA311759397 Enamine

N-(4-aminophenyl)-3-(dimethylamino)propanamide

  • MDL number MFCD09734880
  • Cas Number : 854813-77-7
  • Molecular Weight : 207.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-57842

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57842
Availability details for
Quantity
  • Availability
  • Add to Cart