ENA311759398 Enamine

6,7-dichloro-2-methyl-2,3-dihydro-1-benzofuran-3-one

  • MDL number MFCD16089032
  • Cas Number : 1247582-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-70342

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70342
Availability details for
Quantity
  • Availability
  • Add to Cart