ENA311759399 Enamine

3-[(4-methylphenyl)formamido]-3-phenylpropanoic acid

  • MDL number MFCD00563403
  • Cas Number : 452347-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190...192
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70343

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70343
Availability details for
Quantity
  • Availability
  • Add to Cart