ENA311759401 Enamine

2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)acetonitrile

  • MDL number MFCD12766035
  • Cas Number : 1184639-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70345

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70345
Availability details for
Quantity
  • Availability
  • Add to Cart