ENA311759402 Enamine

3-(4-fluorophenyl)-1-(piperidin-4-yl)urea

  • MDL number MFCD08059705
  • Cas Number : 283167-30-6
  • Molecular Weight : 237.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-57978

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57978
Availability details for
Quantity
  • Availability
  • Add to Cart