ENA311759405 Enamine

1-(6-bromo-1H-indol-3-yl)ethan-1-one

  • MDL number MFCD09834653
  • Cas Number : 316181-82-5
  • Molecular Weight : 238.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 240-242
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70351

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70351
Availability details for
Quantity
  • Availability
  • Add to Cart