ENA311759407 Enamine

(1R,4R)-2-methyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide

  • MDL number MFCD08282791
  • Cas Number : 125224-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70353

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70353
Availability details for
Quantity
  • Availability
  • Add to Cart