ENA311759409 Enamine

2-(2-methyl-2,3-dihydro-1H-indol-1-yl)ethan-1-amine

  • MDL number MFCD09807114
  • Cas Number : 1016522-32-9
  • Molecular Weight : 176.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-58028

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58028
Availability details for
Quantity
  • Availability
  • Add to Cart