ENA311759415 Enamine

3-(5-methylthiophen-2-yl)-3-oxopropanenitrile

  • MDL number MFCD09931289
  • Cas Number : 71683-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105...107
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70367

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70367
Availability details for
Quantity
  • Availability
  • Add to Cart