ENA311759421 Enamine

6-bromo-4-(methylamino)-3,4-dihydro-2H-1lambda6-benzothiopyran-1,1-dione

  • MDL number MFCD12133582
  • Cas Number : 1156176-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70376

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70376
Availability details for
Quantity
  • Availability
  • Add to Cart