ENA311759425 Enamine

N-(1-cyanocyclohexyl)cyclopropanecarboxamide

  • MDL number MFCD12783713
  • Cas Number : 1251240-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-70380

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70380
Availability details for
Quantity
  • Availability
  • Add to Cart