ENA311759426 Enamine

1-methyl-4-methylidenepiperidine

  • MDL number MFCD09834620
  • Cas Number : 13669-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-70381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70381
Availability details for
Quantity
  • Availability
  • Add to Cart