ENA311759433 Enamine

4-(methylamino)benzene-1-sulfonamide

  • MDL number MFCD11207384
  • Cas Number : 16891-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166-167
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58198

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58198
Availability details for
Quantity
  • Availability
  • Add to Cart