ENA311759438 Enamine

4-{[3-(pyrrolidin-1-yl)propyl]sulfamoyl}benzoic acid hydrochloride

  • MDL number MFCD17977055
  • Cas Number : 1203244-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-70415

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70415
Availability details for
Quantity
  • Availability
  • Add to Cart