ENA311759444 Enamine

N-[(1,3-dioxaindan-5-yl)methyl]aniline

  • MDL number MFCD01468475
  • Cas Number : 3526-42-9
  • Molecular Weight : 227.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70428

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70428
Availability details for
Quantity
  • Availability
  • Add to Cart