ENA311759449 Enamine

2-phenyl-1,4-oxazepane

  • MDL number MFCD12183813
  • Cas Number : 933734-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70435

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70435
Availability details for
Quantity
  • Availability
  • Add to Cart