ENA311759450 Enamine

4-fluoro-3-(1H-pyrrol-1-yl)aniline

  • MDL number MFCD12165077
  • Cas Number : 1178165-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70439

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70439
Availability details for
Quantity
  • Availability
  • Add to Cart