ENA311759453 Enamine

1-(2-methoxyethyl)piperidin-3-amine

  • MDL number MFCD14602247
  • Cas Number : 1250372-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70443

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70443
Availability details for
Quantity
  • Availability
  • Add to Cart