ENA311759454 Enamine

2-hydroxy-N-(3-phenylpropyl)benzamide

  • MDL number MFCD01428950
  • Cas Number : 153810-66-3
  • Molecular Weight : 255.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70444
Availability details for
Quantity
  • Availability
  • Add to Cart