ENA311759457 Enamine

4-chloro-5,6-dimethyl-2-(pyridin-2-yl)pyrimidine

  • MDL number MFCD12775388
  • Cas Number : 204394-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70449

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70449
Availability details for
Quantity
  • Availability
  • Add to Cart