ENA311759462 Enamine

methyl 3,5-dimethyl-1H-pyrazole-4-carboxylate

  • MDL number MFCD01846186
  • Cas Number : 25016-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116...118
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70456

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70456
Availability details for
Quantity
  • Availability
  • Add to Cart