ENA311759463 Enamine

N-[(1,3-dioxaindan-5-yl)methyl]-4-fluoroaniline

  • MDL number MFCD00022957
  • Cas Number : 202599-73-3
  • Molecular Weight : 245.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70457
Availability details for
Quantity
  • Availability
  • Add to Cart