ENA311759469 Enamine

1-(3-bromophenyl)-5-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD12134477
  • Cas Number : 1154937-57-1
  • Molecular Weight : 281.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70466

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70466
Availability details for
Quantity
  • Availability
  • Add to Cart