ENA311759477 Enamine

(4-chloro-2-fluorophenyl)(3-methylthiophen-2-yl)methanone

  • MDL number MFCD16057654
  • Cas Number : 1249924-08-0
  • Molecular Weight : 254.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70475
Availability details for
Quantity
  • Availability
  • Add to Cart