ENA311759486 Enamine

4-(1H-imidazol-4-yl)pyridine

  • MDL number MFCD08668577
  • Cas Number : 51746-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189-191
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70486

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70486
Availability details for
Quantity
  • Availability
  • Add to Cart