ENA311759509 Enamine

(3-methylbutan-2-yl)(2,2,2-trifluoroethyl)amine

  • MDL number MFCD09947594
  • Cas Number : 1016878-96-8
  • Molecular Weight : 169.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70513

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70513
Availability details for
Quantity
  • Availability
  • Add to Cart