ENA311759512 Enamine

1-[(1-methyl-1H-imidazol-2-yl)methyl]piperazine

  • MDL number MFCD05864493
  • Cas Number : 880361-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70520
Availability details for
Quantity
  • Availability
  • Add to Cart