ENA311759515 Enamine

(2-ethoxyethyl)(ethyl)amine

  • MDL number MFCD04035011
  • Cas Number : 38256-95-0
  • Molecular Weight : 117.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60508

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60508
Availability details for
Quantity
  • Availability
  • Add to Cart