ENA311759519 Enamine

3-[(4-aminophenyl)sulfanyl]propan-1-ol

  • MDL number MFCD11635129
  • Cas Number : 602306-71-8
  • Molecular Weight : 183.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70528

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70528
Availability details for
Quantity
  • Availability
  • Add to Cart