ENA311759525 Enamine

tert-butyl 3-(2-aminoethyl)pyrrolidine-1-carboxylate

  • MDL number MFCD09759135
  • Cas Number : 1048920-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70538

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70538
Availability details for
Quantity
  • Availability
  • Add to Cart