ENA311759529 Enamine

4-(propan-2-yl)cyclohexan-1-one

  • MDL number MFCD00043479
  • Cas Number : 5432-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Flash Point 80
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61099

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-61099
pictograms GHS02,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart