ENA311759540 Enamine

4-[1-(morpholin-4-yl)ethyl]aniline

  • MDL number MFCD11593291
  • Cas Number : 700804-06-4
  • Molecular Weight : 206.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70566
Availability details for
Quantity
  • Availability
  • Add to Cart