ENA311759543 Enamine

2-(4-amino-1H-pyrazol-1-yl)-N-[4-(methylsulfanyl)phenyl]acetamide

  • MDL number MFCD11980252
  • Cas Number : 1154156-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70573

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70573
Availability details for
Quantity
  • Availability
  • Add to Cart