ENA311759548 Enamine

2-(3,4-dimethoxyphenyl)-2-methylpropan-1-amine

  • MDL number MFCD09881831
  • Cas Number : 23023-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70578
Availability details for
Quantity
  • Availability
  • Add to Cart