ENA311759551 Enamine

2-chloro-5H,6H,7H-cyclopenta[b]pyridine-3-carboxamide

  • MDL number MFCD09864316
  • Cas Number : 183871-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70585

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70585
Availability details for
Quantity
  • Availability
  • Add to Cart