ENA311759555 Enamine

2-(chloromethyl)-5-methylimidazo[1,2-a]pyridine hydrochloride

  • MDL number MFCD04505409
  • Cas Number : 502841-51-2
  • Molecular Weight : 217.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70592

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70592
Availability details for
Quantity
  • Availability
  • Add to Cart