ENA311759556 Enamine

3,4-dihydro-2H-pyran-5-carbonitrile

  • MDL number MFCD17977086
  • Cas Number : 21220-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70593

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70593
Availability details for
Quantity
  • Availability
  • Add to Cart