ENA311759558 Enamine

methyl 3-aminothieno[2,3-b]pyridine-2-carboxylate

  • MDL number MFCD00174679
  • Cas Number : 111042-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191...193
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70599

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70599
Availability details for
Quantity
  • Availability
  • Add to Cart