ENA311759564 Enamine

2-(chloromethyl)-5-(3-methylfuran-2-yl)-1,3,4-oxadiazole

  • MDL number MFCD09052055
  • Cas Number : 937620-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70606

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70606
Availability details for
Quantity
  • Availability
  • Add to Cart